Merck
  • Biodegradation of propargite by Pseudomonas putida, isolated from tea rhizosphere.
Biodegradation of propargite by Pseudomonas putida, isolated from tea rhizosphere.
Journal of hazardous materials (2009-11-07)
Soumik Sarkar, Subbiah Seenivasan, Robert Premkumar Samuel Asir
ABSTRACT
Biodegradation of miticide propargite was carried out in vitro by selected Pseudomonas strains isolated from tea rhizosphere. A total number of 13 strains were isolated and further screened based on their tolerance level to different concentrations of propargite. Five best strains were selected and further tested for their nutritional requirements. Among the different carbon sources tested glucose exhibited the highest growth promoting capacity and among nitrogen sources ammonium nitrate supported the growth to the maximum. The five selected Pseudomonas strain exhibited a range of degradation capabilities. Mineral salts medium (MSM) amended with glucose provided better environment for degradation with the highest degradation potential in strain SPR 13 followed by SPR 8 (71.9% and 69.0% respectively).