Merck
  • Temperature-controlled ionic liquid-dispersive liquid-phase microextraction for preconcentration of chlorotoluron, diethofencarb and chlorbenzuron in water samples.
Temperature-controlled ionic liquid-dispersive liquid-phase microextraction for preconcentration of chlorotoluron, diethofencarb and chlorbenzuron in water samples.
Journal of separation science (2009-10-31)
Qingxiang Zhou, Xiaoguo Zhang, Guohong Xie, Junping Xiao
ABSTRACT
This article describes a new, rapid and sensitive method for the determination of chlorotoluron, diethofencarb and chlorbenzuron from water samples with temperature-controlled ionic liquid-dispersive liquid-phase microextraction. In the preconcentration procedure, ionic liquid 1-hexyl-3-methylimidazolium hexafluorophosphate [C(6)MIM] [PF(6)] was employed as the extraction solvent. The parameters, such as volume of [C(6)MIM] [PF(6)], sample pH, extraction time, centrifuging time, temperature and salting-out effect, were investigated in detail. Under the optimal extraction conditions, it has been found that three analytes had excellent LODs (S/N=3) in the range of 0.04-0.43 microg/L. The RSDs (n=6) were in the range of 1.3-4.7%. The proposed method was evaluated with lake water, tap water and melted snow water samples. The experimental results indicated that the proposed method had excellent prospect and would be widely used in the future.
Product Description
Supelco
Diethofencarb, PESTANAL®, analytical standard
Supelco
Chlortoluron, PESTANAL®, analytical standard