Merck
  • Prefluorescent nitroxide probe for the highly sensitive determination of peroxyl and other radical oxidants.
Prefluorescent nitroxide probe for the highly sensitive determination of peroxyl and other radical oxidants.
Analytical chemistry (2009-10-01)
Min Jia, Yu Tang, Yiu-Fai Lam, Sarah A Green, Neil V Blough
ABSTRACT
Fluorescamine derivatized 3-amino-2,2,5,5,-tetramethyl-1-pyrrolidinyloxy (I) is shown to undergo an irreversible reaction with peroxyl radicals and other radical oxidants to generate a more highly fluorescent diamagnetic product (II) and thus can be used as a highly sensitive and versatile probe to determine oxidant production optically, either by monitoring the changes in fluorescence intensity, by HPLC analysis with fluorescence detection, or by a combination of both approaches. By changing the [O2]/[I] ratio, we show that peroxyl radicals can be detected and quantified preferentially in the presence of other radical oxidants. Detection of photochemically produced peroxyl radicals is achieved by employing 3-amino-2,2,5,5,-tetramethyl-1-pyrrolidinyloxy (3-ap) alone, followed by derivatization with fluorescamine. With employment of HPLC analysis, the detection limit of II at a S/N of 2 is approximately 3 nM for a 125 microL injection. Preliminary applications include the detection of peroxyl radicals generated thermally in soybean phosphatidylcholine liposomes and produced photochemically in tap water.
Sigma-Aldrich
Fluorescamine, ≥98% (TLC), powder, used for detection of primary amines