Merck
Cation binding of antimicrobial sulfathiazole to leonardite humic acid.
Environmental science & technology (2009-09-22)
Merle K Richter, Michael Sander, Martin Krauss, Iso Christl, Manuel G Dahinden, Manuel K Schneider, René P Schwarzenbach
ABSTRACT
Sorption of sulfathiazole (STA) and three structural analogs to Leonardite humic acid (LHA) was investigated in single- and binary-solute systems to elucidate the sorption mechanism of sulfonamides to soil organic matter (SOM). Cation binding of STA+ to anionic sites A- in LHA governed sorption up to circumneutral pH, based on the following findings: (i) From pH 7.7 to 3.3, the increase in extent and nonlinearity (i.e., concentration dependence) of STA sorption paralleled the increase in STA+. (ii) From pH 3.3 to 1.7, sorption decreased and nonlinearity increased, consistent with strong competition of STA+ and H+ for A-. (iii) Replacement of the protonable aniline group in STA by an apolar methylbenzene group resulted in much weaker, linear, and pH-independent sorption. (iv) Only analogs with aniline moieties displaced STA from LHA in binary-solute systems. Displacement occurred up to pH 5.4, at which <1% of STA in solution was cationic. (v) STA sorption was well-described (R2 = 0.98) by the NICA-Donnan cation-binding model, yielding high median affinities for STA+ to carboxylic and phenolic A- (log K(STA+,1) = 3.25 +/- 0.08 log (L mol(-1)) and log K(STA+,2) = 8.76 +/- 0.11 log (L mol(-1)), respectively). High affinity cation binding explains sorption of polar sulfonamides in agricultural soils and the strong dependence of sorption on SOM content and pH.
Product Description
Supelco
Sulfathiazole, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Sulfathiazole
Pricing and availability is not currently available.