Merck
  • Use of 1,3-dithiane combined with aryldiazonium cation for immobilization of biomolecules based on electrochemical addressing.
Use of 1,3-dithiane combined with aryldiazonium cation for immobilization of biomolecules based on electrochemical addressing.
Chemical communications (Cambridge, England) (2009-08-05)
Al-Monsur Jiaul Haque, Seung-Ryong Kwon, Hyejin Park, Tae-Hyun Kim, Young-Seok Oh, Sung-Yool Choi, Jong-Dal Hong, Kyuwon Kim
ABSTRACT
We report the use of 1,3-dithiane combined with aryldiazonium cation for the immobilization of biomolecules based on electrochemical addressing.
Sigma-Aldrich
1,3-Dithiane, 97%
Pricing and availability is not currently available.