Merck
  • Crystal structure of a phospholipase A(2) homolog complexed with p-bromophenacyl bromide reveals important structural changes associated with the inhibition of myotoxic activity.
Crystal structure of a phospholipase A(2) homolog complexed with p-bromophenacyl bromide reveals important structural changes associated with the inhibition of myotoxic activity.
Biochimica et biophysica acta (2009-07-21)
Daniela P Marchi-Salvador, Carlos A H Fernandes, Lucas B Silveira, Andreimar M Soares, Marcos R M Fontes
ABSTRACT
For the first time, the structure of a catalytic inactive phospholipase A(2) homolog (Lys49-PLA(2)s) complexed with p-bromophenacyl bromide (BPB) has been solved by X-ray crystallography. Lys49-PLA(2)s are among the main components of Viperidae snake venoms, causing myonecrosis and other actions despite their catalytic inactivity. BPB, a classic inhibitor of catalytic-active PLA(2)s, has been used since the 1970s because it binds specifically the His48 residue of the catalytic site. Curiously, when Lys49-PLA(2) is chemically modified by BPB, it causes a partial inhibition of the myotoxic function which is associated with the C-terminus and not with the catalytic site. The structure of PrTX-I complexed to BPB revealed unambiguously that the inhibitor binds covalently to His48, causing a distortion of the Ca(2)(+)-binding loop region and C-terminus rearrangement in one of its monomers. The comparison between the apo and BPB-complexed PrTX-I structures showed an increased symmetry between the two monomers with the formation of an interchain hydrogen bond between Tyr119 residues. PrTX-I undergoes tertiary and quaternary structural changes when complexed to BPB which could be related to reduction of myotoxicity and other toxic activities. We also proposed a novel myotoxic inhibition hypothesis integrating "myotoxic" and "active" sites for bothropic Lys49-PLA(2)s.
Sigma-Aldrich
2,4′-Dibromoacetophenone, >98%
Pricing and availability is not currently available.
Supelco
2,4′-Dibromoacetophenone, for HPLC derivatization, LiChropur, ≥99.0% (HPLC)
Pricing and availability is not currently available.