Merck
  • Estradiol-17beta and linseed meal interact to alter visceral organ mass and hormone concentrations from ovariectomized ewes.
Estradiol-17beta and linseed meal interact to alter visceral organ mass and hormone concentrations from ovariectomized ewes.
Domestic animal endocrinology (2009-06-30)
M R O'Neil, G P Lardy, M E Wilson, C O Lemley, L P Reynolds, J S Caton, K A Vonnahme
ABSTRACT
To evaluate the estrogenic potential of secoisolariciresinol diglycoside (SDG) found in linseed meal (LSM) on visceral organ mass, IGF-I, and thyroid hormone (T(3) and T(4)) concentrations, 48 multiparous, ovariectomized ewes (54.6 +/- 1.1 kg) were used in a 3 x 4 factorial arrangement. Main effects were length of LSM feeding (0, 1, 7, or 14 d) and length of exposure to estradiol-17beta (E(2)) implant (0, 6, or 24 h prior to tissue collection). Implanting ewes with E(2) for 24h increased liver mass relative to empty body weight (EBW; g/kg EBW) compared with ewes implanted for 0 or 6 h (P <or= 0.03), whereas feeding LSM for 14 d decreased liver mass compared with ewes fed LSM for 1 or 7 d (P <or= 0.02). There was an LSM x E(2) interaction (P = 0.01) for duodenal mass (g/kg EBW), LSM, and E(2) tended (P = 0.07) to influence the stomach complex mass; however, ileal mass was not affected. Neither LSM nor E(2) affected (P >or= 0.12) CYP2C or CYP3A mRNA expression or cellularity of the liver. Exogenous E(2) influenced circulating concentrations of IGF-I, T(3), and T(4). The estrogenic or anti-estrogenic potential of LSM is dependent upon the tissue, exposure to E(2), and the duration of LSM feeding. Feeding LSM during gestation, lactation, or during the grow-finish phase warrants further investigation.
Sigma-Aldrich
Secoisolariciresinol, ≥95.0% (HPLC)