Merck
  • Deprotection of the indole (N(ind))-formyl (For) group on tryptophan employing a new reagent, N,N'-dimethylethylendiamine (DMEDA) in an aqueous solution.
Deprotection of the indole (N(ind))-formyl (For) group on tryptophan employing a new reagent, N,N'-dimethylethylendiamine (DMEDA) in an aqueous solution.
Chemical & pharmaceutical bulletin (2009-02-03)
Takenao Odagami, Yuko Tsuda, Yuji Kogami, Hiroyuki Kouji, Yoshio Okada
ABSTRACT
The deprotection of the indole (N(ind))-formyl (For) group on Trp was achieved in a 95% yield using N,N'-dimethylethylendiamine (DMEDA) (1.5, 2.0, 3.0 eq) in water at room temperature. A new reagent was successfully applied to the deprotection of a model peptide, H-Phe-Trp(N(ind)-For)-Lys-Tyr-OH, to give H-Phe-Trp-Lys-Tyr-OH in a 91% yield.
Sigma-Aldrich
N,N′-Dimethylethylenediamine, 98%
Sigma-Aldrich
N,N′-Dimethylethylenediamine, 85%