Merck
  • Knockdown of TRPC3 with siRNA coupled to carbon nanotubes results in decreased insulin-mediated glucose uptake in adult skeletal muscle cells.
Knockdown of TRPC3 with siRNA coupled to carbon nanotubes results in decreased insulin-mediated glucose uptake in adult skeletal muscle cells.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2009-01-15)
Johanna T Lanner, Joseph D Bruton, Yohannes Assefaw-Redda, Zoita Andronache, Shi-Jin Zhang, Denise Severa, Zhi-Bin Zhang, Werner Melzer, Shi-Li Zhang, Abram Katz, Håkan Westerblad
ABSTRACT
The involvement of Ca(2+) in the insulin-mediated signaling cascade, resulting in glucose uptake in skeletal muscle, is uncertain. Here, we test the hypothesis that Ca(2+) influx through canonical transient receptor potential 3 (TRPC3) channels modulates insulin-mediated glucose uptake in adult skeletal muscle. Experiments were performed on adult skeletal muscle cells of wild-type (WT) and obese, insulin-resistant ob/ob mice. Application of the diacylglycerol analog 1-oleyl-2-acetyl-sn-glycerol (OAG) induced a nonselective cation current, which was inhibited by the addition of anti-TRPC3 antibody in the patch pipette and smaller in ob/ob than in WT cells. Knockdown of TRPC3, using a novel technique based on small interfering RNA (siRNA) coupled to functionalized carbon nanotubes, resulted in pronounced (approximately 70%) decreases in OAG-induced Ca(2+) influx and insulin-mediated glucose uptake. TRPC3 and the insulin-sensitive glucose transporter 4 (GLUT4) coimmunoprecipitated, and immunofluorescence staining showed that they were colocalized in the proximity of the transverse tubular system, which is the predominant site of insulin-mediated glucose transport in skeletal muscle. In conclusion, our results indicate that TRPC3 interacts functionally and physically with GLUT4, and Ca(2+) influx through TRPC3 modulates insulin-mediated glucose uptake. Thus, TRPC3 is a potential target for treatment of insulin-resistant conditions.
Sigma-Aldrich
1-Oleoyl-2-acetyl-sn-glycerol, ≥97% (TLC), oil