Merck
  • Detection of the superoxide radical anion using various alkanethiol monolayers and immobilized cytochrome c.
Detection of the superoxide radical anion using various alkanethiol monolayers and immobilized cytochrome c.
Analytical chemistry (2008-12-17)
Xiaojun J Chen, Alan C West, Donald M Cropek, Scott Banta
ABSTRACT
The superoxide radical anion (SO) is a critical biomarker for monitoring cellular stress responses. Electrochemical SO biosensors are frequently constructed through the covalent immobilization of cytochrome c (Cyt c) onto self-assembled monolayers (SAMs); however, a detailed comparison of these systems as well as configuration influence on SO detection is needed to enable robust applications. Two reaction pathways, oxidation of SO by the SAM-modified gold electrode or electron transfer through a protein and monolayer relay, may be involved during the electrochemical detection of SO with Cyt c, depending on the SAM that is used. Although electrodes with SAMs alone can exhibit a high sensitivity and low limit of detection (LOD) for the SO, they can suffer from a strong response to the presence of interferents such as hydrogen peroxide and ascorbic acid. Electrodes with immobilized Cyt c show decreased sensitivity, but exhibit better selectivity and resistance to fouling in complex media. Considering the trade-offs between sensitivity, selectivity, and LOD for SO detection, a bioelectrode made with Cyt c immobilized on dithiobis(succinimidyl)propionate (DTSP) appears to be the most suitable configuration. In phosphate buffer, the DTSP/Cyt c electrode has a sensitivity of 410 nA microM(-1) cm(-2) and an LOD for SO of 73 nM. Results are also presented for the detection of SO in a complex tissue culture media (MEM) with and without serum, and the sensitivity of the DTSP/Cyt c in MEM in the absence of serum increased to 640 nA microM(-1) cm(-2). By measuring SO with a DTSP/Cyt c electrode before and after the addition of a bolus of the superoxide dismutase (SOD) enzyme, the specificity of the SOD enzyme can be combined with the sensitivity of Cyt c system.
Sigma-Aldrich
3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester), powder
Pricing and availability is not currently available.