Merck
  • Synthesis of indoles via domino reaction of N-aryl amides and ethyl diazoacetate.
Synthesis of indoles via domino reaction of N-aryl amides and ethyl diazoacetate.
Journal of the American Chemical Society (2008-09-19)
Sun-Liang Cui, Jing Wang, Yan-Guang Wang
ABSTRACT
A general and concise synthesis of functionalized indoles via domino reaction of N-aryl amides and ethyl diazoacetate has been developed. The methodology offers a great potential for the synthesis of biologically active and naturally occurring indole derivatives.
Sigma-Aldrich
Ethyl diazoacetate, contains ≥13 wt. % dichloromethane