- Guiding the self-assembly of a second-generation polyphenylene dendrimer into well-defined patterns.
Guiding the self-assembly of a second-generation polyphenylene dendrimer into well-defined patterns.
A second-generation polyphenylene dendrimer 1 is shown to self-assemble into nanofibers. To guide the formation of the dendrimer fibers into well-defined patterns, 1H,1H,2H,2H-perfluorodecyltrichlorosilane is grafted in the gas phase onto a silicon substrate. De-wetting of the solution on the nanopatterned surface results in the formation of a nanostructured template, into which fiber growth subsequently occurs under the constraints set by the de-wetted morphology.