Merck
  • Epithelial and endothelial barriers in the olfactory region of the nasal cavity of the rat.
Epithelial and endothelial barriers in the olfactory region of the nasal cavity of the rat.
Histochemistry and cell biology (2008-03-15)
Hartwig Wolburg, Karen Wolburg-Buchholz, Heike Sam, Sándor Horvát, Maria A Deli, Andreas F Mack
ABSTRACT
The olfactory ensheathing (glial) cells (OECs) have been identified to be useful candidate cells to support regeneration after being transplanted into injured fiber tracts of the central nervous system. We investigated by means of immunocytochemistry and freeze-fracturing the morphology and molecular composition of OEC tight junctions in the rat olfactory system. In addition, we tested the hypothesis whether tight junctions and orthogonal arrays of particles (OAPs) which contain the water channel protein aquaporin-4 (AQP4), are mutually exclusive as suggested in previous studies. In OECs, we found neither OAPs nor AQP4, but tight junctions immunoreactive for ZO-1, occludin, and claudin-5, but immunonegative for ZO-2 and claudin-3. To shed more light on the function of OEC tight junctions, we tested the permeability and tight junction composition of blood vessels and fila olfactoria. We found them both, permeable for infused lanthanum nitrate, and to be immunopositive for ZO-1 and claudin-5. The tight junctions of the OECs are discussed to be responsible for micro-compartmentalization within the olfactory fiber tract providing a benefit for axonal growth.
Sigma-Aldrich
Lanthanum(III) nitrate hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Lanthanum(III) nitrate hexahydrate, 99.99% trace metals basis