Merck
Fragrance material review on ethyl salicylate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-21)
A Lapczynski, D McGinty, L Jones, S Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicological and dermatological review of ethyl salicylate when used as a fragrance ingredient is presented.
Sigma-Aldrich
Ethyl salicylate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl salicylate, 99%