Merck
  • [FeIII(tmdta)]--twist-boat/half-chair conformer ratio reliably deduced from DFT-calculated Raman spectra.
[FeIII(tmdta)]--twist-boat/half-chair conformer ratio reliably deduced from DFT-calculated Raman spectra.
Chemical communications (Cambridge, England) (2007-09-27)
Roland Meier, Joachim Maigut, Bernd Kallies, Nicolai Lehnert, Florian Paulat, Frank W Heinemann, Gernot Zahn, Martin P Feth, Harald Krautscheid, Rudi van Eldik
ABSTRACT
The equilibrium between the twist-boat (tb) and half-chair (hc) conformers of the central diamine chelate ring of [Fe(III)(tmdta)]- in solids and aqueous solution has been studied by Raman spectroscopy, supported by calculated Raman spectra using Density Functional Theory.
Sigma-Aldrich
1,3-Diaminopropane, ≥99%
Sigma-Aldrich
1,3-Diaminopropane dihydrochloride, 98%