Merck
  • Mechanisms involved in the cytotoxic and cytoprotective actions of saturated versus monounsaturated long-chain fatty acids in pancreatic beta-cells.
Mechanisms involved in the cytotoxic and cytoprotective actions of saturated versus monounsaturated long-chain fatty acids in pancreatic beta-cells.
The Journal of endocrinology (2007-07-21)
Eleftheria Diakogiannaki, Shalinee Dhayal, Caroline E Childs, Philip C Calder, Hannah J Welters, Noel G Morgan
ABSTRACT
Long-chain saturated and monounsaturated fatty acids differ in their propensity to induce beta-cell death in vitro with palmitate (C16:0) being cytotoxic, whereas palmitoleate (C16:1n-7) is cytoprotective. We now show that this cytoprotective capacity extends to a poorly metabolised C16:1n-7 derivative, methyl-palmitoleate (0.25 mM palmitate alone: 92 +/- 4% death after 18 h; palmitate plus 0.25 mM methyl-palmitoleate: 12 +/- 2%; P < 0.001). Palmitoleate and its methylated derivative also acted as mitogens in cultured beta-cells (5-bromo-2-deoxyuridine incorporation - control: 0.15 +/- 0.01 units; 0.25 mM palmitoleate: 0.22 +/- 0.01 units; P < 0.05). It has been proposed that alterations in neutral lipid synthesis (particularly triacylglycerol (TAG) formation) might mediate the differential responses to saturated and unsaturated fatty acids and we have examined this proposition. Palmitate and palmitoleate both promoted beta-cell phospholipid remodelling and increased TAG formation (control: 0.9 +/- 0.1 nmol TAG/10(6) cells; 0.25 mM palmitate: 1.55 +/- 0.07; 0.25 mM palmitoleate: 1.4 +/- 0.05; palmitate plus palmitoleate: 2.3 +/- 0.1). By contrast, methyl-palmitoleate failed to influence TAG levels (0.25 mM methyl-palmitoleate alone: 0.95 +/- 0.06 nmol TAG/10(6) cells; methyl-palmitoleate plus palmitate: 1.5 +/- 0.05) or its fatty acid composition in beta-cells exposed to palmitate. The results suggest that monounsaturated fatty acids can promote cell viability and mitogenesis by a mechanism that does not require their metabolism and is independent of alterations in TAG formation.
Product Description
Supelco
Methyl palmitoleate, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Methyl palmitoleate, ≥99% (capillary GC), liquid
Pricing and availability is not currently available.