Merck
  • Optimizing TiO2-based phosphopeptide enrichment for automated multidimensional liquid chromatography coupled to tandem mass spectrometry.
Optimizing TiO2-based phosphopeptide enrichment for automated multidimensional liquid chromatography coupled to tandem mass spectrometry.
Analytical chemistry (2007-05-26)
Greg T Cantin, Teresa R Shock, Sung Kyu Park, Hiten D Madhani, John R Yates
ABSTRACT
An automated online multidimensional liquid chromatography system coupled to ESI-based tandem mass spectrometry was used to assess the effectiveness of TiO2 in the enrichment of phosphopeptides from tryptic digests of protein mixtures. By monitoring the enrichment of phosphopeptides, an optimized set of loading, wash, and elution conditions were realized for TiO2. A comparison of TiO2 with other resins used for phosphopeptide enrichment, Fe(III)-IMAC and ZrO2, was also carried out using tryptic digests of both simple and moderately complex protein mixtures; where TiO2 was shown to be superior in performance.
Sigma-Aldrich
4-Imidazoleacetic acid hydrochloride, 98%