Merck
Polymorphism and thermodynamics of m-hydroxybenzoic acid.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2006-06-20)
Fredrik L Nordström, Ake C Rasmuson
ABSTRACT
Solution and solid-state properties of m-hydroxybenzoic acid have been investigated. Two polymorphs were found where the monoclinic modification exhibits a higher stability than the orthorhombic form. The solubility of the monoclinic polymorph was determined between 10 and 50 degrees C in methanol, acetonitrile, acetic acid, acetone, water and ethyl acetate. The solubility of the orthorhombic polymorph was determined between 10 and 50 degrees C in acetonitrile, acetic acid, acetone and ethyl acetate. A thermodynamic analysis revealed a marked correlation between the molar solubility and the van't Hoff enthalpy of solution at constant temperature. In addition, in each solvent increased temperature resulted in increased van't Hoff enthalpy of solution. It is shown that the solubility data can be used to estimate melting properties for both polymorphs. The solubility ratio of the two forms and the DSC thermogram of the orthorhombic form strongly suggest that the system is monotropic. However, according to the polymorph rules of Burger and Ramberger, the estimated higher melting enthalpy and lower melting temperature of the orthorhombic form points towards an enantiotropic system. Hence, this system appears to be an exception to the Burger and Ramberger melting enthalpy rule, and the probable reason for this is found in the difference in the heat capacity of the two solid forms.
Sigma-Aldrich
3-Hydroxybenzoic acid, ReagentPlus®, 99%