Merck
  • Protective effects of trapidil in ischemia-reperfusion injury due to testicular torsion and detorsion: an experimental study.
Protective effects of trapidil in ischemia-reperfusion injury due to testicular torsion and detorsion: an experimental study.
International journal of urology : official journal of the Japanese Urological Association (2006-06-15)
Salih Somuncu, Murat Cakmak, Sibel Erdogan, Osman Caglayan, Hülya Akman, Murat Kaya
ABSTRACT
We aimed to detect the preventive effects of trapidil in ischemia-reperfusion (IR) injury due to testicular torsion and detorsion. Forty prepubertal albino rats were used. In the IR group, torsion was created by rotating the left testis over 2 h, and detorsion was done by untwisting the testis. Bilateral orchiectomies were performed after 4 h. In study group, 2-h torsion was performed and trapidil was administered as a single dose 1 h before detorsion. Bilateral orchiectomies were performed after 4 h. In the sham group, a sham operation was done. In the sham plus trapidil group, a sham operation was done and trapidil was administered as a single dose. Testicular tissue malondialdehyde (MDA), nitric oxide (NO) and total sulfhydryl (T-SH) levels were determined for each group. The grades of interstitial injury were determined in histopathologic examination. The NO and MDA levels in the IR group were significantly higher than the study, sham and sham plus trapidil groups in the left testis (P<0.05, P<0.001 and P<0.001, respectively). A statistical difference was not found among study, sham and sham plus trapidil groups in the left testis in NO and MDA levels (P>0.05). The T-SH level in the study group was significantly higher than in the IR, sham and sham plus trapidil groups in left testis P<0.05). In the IR group (left testis), grade 1 interstitial injury was 30% (3/10), grade 2 injury was 60% (6/10) and grade 3 injury was 10% (1/10). In the study group (left testis), grade 1 interstitial injury was 30% (3/10) and there was no injury in 70% (7/10). Trapidil decreased free oxygen radical formation in testicular torsion and detorsion, and attenuated histopathological damage in the ipsilateral twisted testis.
Product Description
Trapidil, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.