- [Synthesis of new test reagents for the estrogen receptor in human breast cancer tissues].
[Synthesis of new test reagents for the estrogen receptor in human breast cancer tissues].
Yao xue xue bao = Acta pharmaceutica Sinica (1991-01-01)
F Mao, L L Weng, H Zheng
PMID1666730
ABSTRACT
According to the character of structure and activity of the test reagents I-IV, two new reagents V and VI for detecting estrogen receptor of human mammary cancer cells were synthesized. This simplifies the route of synthesis and increases activity. Key intermediates VIII and IX were confirmed by IR, MS, UV and elemental analysis. The quantification of the final products V and VI were determined by UV. The result of preliminary clinico-pathological test shows compound V to be effective on estrogen receptor.
MATERIALS