- Determination of free amino acid enantiomers in rat brain and serum by high-performance liquid chromatography after derivatization with N-tert.-butyloxycarbonyl-L-cysteine and o-phthaldialdehyde.
Determination of free amino acid enantiomers in rat brain and serum by high-performance liquid chromatography after derivatization with N-tert.-butyloxycarbonyl-L-cysteine and o-phthaldialdehyde.
Journal of chromatography (1992-11-06)
A Hashimoto, T Nishikawa, T Oka, K Takahashi, T Hayashi
PMID1491056
ABSTRACT
The concurrent determination of free amino acid enantiomers and non-chiral amino acids in rat brain and serum was accomplished by high-performance liquid chromatography with fluorimetric detection after derivatization with N-tert.-butyloxycarbonyl-L-cysteine and o-phthaldialdehyde. The method revealed the presence of a large amount of free D-serine (0.22 mumol/g of tissue; D/D + L ratio = 0.25) in the brain whereas D-aspartate and D-alanine were established to be at trace levels. These results further support the presence of D-serine in adult brain tissues as demonstrated by recent work using gas chromatography.