Merck
  • Efficient glycosidation of a phenyl thiosialoside donor with diphenyl sulfoxide and triflic anhydride in dichloromethane.
Efficient glycosidation of a phenyl thiosialoside donor with diphenyl sulfoxide and triflic anhydride in dichloromethane.
Organic letters (2006-02-24)
David Crich, Wenju Li
ABSTRACT
The formation of sialic acid glycosides with a thiosialic acid derivative, diphenyl sulfoxide, and trifluoromethanesulfonic anhydride is reported. With an excess of diphenyl sulfoxide, glycal formation can be completely suppressed and excellent yields are obtained for coupling to a wide range of primary, secondary, and tertiary acceptors.
Sigma-Aldrich
Diphenyl sulfoxide, 96%