Merck
  • Analysis of residual monomers in dendritic methacrylate copolymers and composites by HPLC and headspace-GC/MS.
Analysis of residual monomers in dendritic methacrylate copolymers and composites by HPLC and headspace-GC/MS.
Dental materials : official publication of the Academy of Dental Materials (2005-12-29)
Eeva K Viljanen, Sarka Langer, Mikael Skrifvars, Pekka K Vallittu
ABSTRACT
The aim of this study was to analyze the residual monomer content of photopolymerized dendritic methacrylate copolymers and particulate filler composites. Headspace-gas chromatography/mass spectrometry (HS-GC/MS) was compared with high performance liquid chromatography (HPLC). The resin mixtures consisted of a dendritic methacrylate monomer, methyl methacrylate and acetoacetoxyethyl methacrylate in varied proportions. In addition, one of the composites contained 1,4-butanediol dimethacrylate. Camphorquinone and 2-(N,N-dimethylamino)ethyl methacrylate were used as the light-activated initiator system. The content of residual methyl methacrylate and acetoacetoxyethyl methacrylate after 40 s photopolymerization were analyzed with HPLC and HS-GC/MS. The content of residual methyl methacrylate decreased and residual acetoacetoxyethyl methacrylate increased with increasing concentration of acetoacetoxyethyl methacrylate in the resin mixture. The results with both methods had the same trend. The addition of acetoacetoxyethyl methacrylate enhanced the copolymerization of methyl methacrylate, but did not decrease the total residual monomer content. The HS-GC/MS method was found to be a feasible method in the analysis of low-boiling residuals in dental polymers.
Sigma-Aldrich
1,4-Butanediol dimethacrylate, 95%, contains 200-300 ppm MEHQ as inhibitor
Pricing and availability is not currently available.