Merck
Acute barium nitrate intoxication treated by hemodialysis.
Acta anaesthesiologica Scandinavica (2005-01-29)
H Bahlmann, R Lindwall, H Persson
ABSTRACT
A 22-year-old male was admitted to hospital with diarrhea and vomiting, cardiac arrhythmias, severe hypokalemia and gradual onset of muscular weakness. A potassium infusion was started, but for several hours serum potassium remained low. Evidence of toxic ingestion was initially lacking. When it became clear -- after a considerable delay -- that the patient had ingested barium nitrate, hemodialysis was started. This resulted in rapid clinical improvement with correction of hypokalemia and restored muscular function. Intoxication with barium causes hypokalemia, arrhythmias, muscular weakness and paralysis, often requiring respiratory support. This patient presented with symptoms typical of severe barium intoxication, non-responsive to potassium supplementation. There are few published reports on the use of hemodialysis in barium poisoning. This case confirms the possible benefit of hemodialysis in severe cases, where potassium supplementation alone is insufficient.
Sigma-Aldrich
Barium nitrate, ACS reagent, ≥99%
Sigma-Aldrich
Barium nitrate, puriss. p.a., ACS reagent, ≥99%, ≥52% Ba basis
Sigma-Aldrich
Barium nitrate, 99.999% trace metals basis