Merck
  • Determination of diethanolamine or N-methyldiethanolamine in high ammonium concentration matrices by capillary electrophoresis with indirect UV detection: application to the analysis of refinery process waters.
Determination of diethanolamine or N-methyldiethanolamine in high ammonium concentration matrices by capillary electrophoresis with indirect UV detection: application to the analysis of refinery process waters.
Analytical and bioanalytical chemistry (2004-09-01)
N Bord, G Crétier, J-L Rocca, C Bailly, J-P Souchez
ABSTRACT
Alkanolamines such as diethanolamine (DEA) and N-methyldiethanolamine (MDEA) are used in desulfurization processes in crude oil refineries. These compounds may be found in process waters following an accidental contamination. The analysis of alkanolamines in refinery process waters is very difficult due to the high ammonium concentration of the samples. This paper describes a method for the determination of DEA in high ammonium concentration refinery process waters by using capillary electrophoresis (CE) with indirect UV detection. The same method can be used for the determination of MDEA. Best results were achieved with a background electrolyte (BGE) comprising 10 mM histidine adjusted to pH 5.0 with acetic acid. The development of this electrolyte and the analytical performances are discussed. The quantification was performed by using internal standardization, by which triethanolamine (TEA) was used as internal standard. A matrix effect due to the high ammonium content has been highlighted and standard addition was therefore used. The developed method was characterized in terms of repeatability of migration times and corrected peak areas, linearity, and accuracy. Limits of detection (LODs) and quantification (LOQs) obtained were 0.2 and 0.7 ppm, respectively. The CE method was applied to the determination of DEA or MDEA in refinery process waters spiked with known amounts of analytes and it gave excellent results, since uncertainties obtained were 8 and 5%, respectively.
Sigma-Aldrich
N-Methyldiethanolamine, ≥99%
Pricing and availability is not currently available.