Merck
Constellational diastereomers in encapsulation complexes.
Chemical communications (Cambridge, England) (2004-07-28)
Masamichi Yamanaka, Julius Rebek
ABSTRACT
Three chiral guests in a cylindrical capsule led to six diastereomeric complexes.