Merck
Total synthesis of natural (+)-(2's,3'r)-zoapatanol.
Organic letters (2004-06-18)
Catherine Taillier, Véronique Bellosta, Janine Cossy
ABSTRACT
[reaction: see text] (+)-Zoapatanol was synthesized by using four key-steps: a Suzuki cross-coupling to prepare a (Z)-alpha,beta-unsaturated ester followed by an enantioselective dihydroxylation to control the C2' and C3' stereocenters, an intramolecular Horner-Wadsworth-Emmons olefination to construct the oxepane ring, and a chemoselective nucleophilic addition/Birch reduction process of a Weinreb amide to introduce simultaneously the beta,gamma-unsaturated ketone on the side-chain and regenerate alcohols from benzyl ethers.
Sigma-Aldrich
Bromotrimethylsilane, 97%
Sigma-Aldrich
Bromotrimethylsilane, purum, ≥97.0% (AT)