Merck
  • Technological parameters of the ammonolysis of waste 1,2-dichloropropane.
Technological parameters of the ammonolysis of waste 1,2-dichloropropane.
Journal of hazardous materials (2004-06-05)
Marcin Bartkowiak, Grzegorz Lewandowski, Eugeniusz Milchert
ABSTRACT
The ammonolysis of waste 1,2-dichloropropane (DCP) using liquid ammonia has been investigated. The influence of temperature, molar ratio of NH3/1,2-dichloropropane and reaction time was examined. The highest yield of the synthesis was achieved at a temperature of 140 degrees C, for the reaction time of 3 h and the molar ratio of NH3/1,2-dichloropropane as 20:1. Under these conditions the degree of 1,2-dichloropropane conversion amounted to 97.1 mol% and the selectivity of transformation to 1,2-diaminopropane (DAP) in relation to consumed 1,2-dichloropropane was 25.3 mol%, whereas in relation to consumed ammonia 17.9 mol%. The remaining 1,2-dichloropropane reacts to form polypropyleneamines.
Product Description
Supelco
1,2-Dichloropropane, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
1,2-Dichloropropane, 99%
Pricing and availability is not currently available.