Merck
  • Developmental changes in glutathione S-transferase isoforms expression and activity in intrasplenic fetal liver tissue transplants in rats.
Developmental changes in glutathione S-transferase isoforms expression and activity in intrasplenic fetal liver tissue transplants in rats.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie (2003-11-19)
Amelie Lupp, Tino Anschütz, Pirjo Lindström-Seppä, Dieter Müller
ABSTRACT
The aim of the present study was to characterise developmental changes in glutathione S-transferase (GST) isoforms expression and in glutathione conjugation capacity in intrasplenic liver tissue transplants. For this purpose, syngenic fetal liver tissue suspensions were transplanted into the spleens of adult male Fischer 344 rats. Three days, 1, 2, 4 weeks, 2, 4, 6 months and 1 year later, transplant-recipients and control animals were sacrificed and class alpha, mu and pi GST isoforms expression and GST activities using the substrates o-dinitrobenzene and 1-chloro-2,4-dinitrobenzene were assessed in livers and spleens. In the hepatocytes of the adult livers no class pi, but a distinct class alpha and mu GST expression was seen. The bile duct epithelia were class pi GST positive. Fetal livers displayed almost no class alpha and mu, but a slight class pi GST expression. The same pattern was seen in 3-day-old intrasplenic liver tissue transplants. Up to 2 weeks after surgery the class alpha and mu GST expression increased in the hepatocytes of the transplants, whereas the immunostaining for class pi GST disappeared. No remarkable changes were seen thereafter. Normal conjugation capacities were observed with the livers of both groups of rats. Control spleens displayed only low GST activities. From 2 months after transplantation on activities were significantly higher in transplant-containing spleens than in respective control organs with a further increase up to one year after grafting. These results show that intrasplenically transplanted fetal liver cells proliferate and differentiate into mature cells displaying a GST expression pattern with respective enzyme activities similar to adult liver.
Sigma-Aldrich
1,2-Dinitrobenzene, 97%