Merck
  • Efficient synthesis of glycosylated phenazine natural products and analogs with DISAL (methyl 3,5-dinitrosalicylate) glycosyl donors.
Efficient synthesis of glycosylated phenazine natural products and analogs with DISAL (methyl 3,5-dinitrosalicylate) glycosyl donors.
Organic & biomolecular chemistry (2003-10-07)
Jane B Laursen, Lars Petersen, Knud J Jensen, John Nielsen
ABSTRACT
Inspired by the occurrence and function of phenazines in natural products, new glycosylated analogs were designed and synthesized. DISAL (methyl 3,5-dinitrosalicylate) glycosyl donors were used in an efficient and easily-handled glycosylation protocol compatible with combinatorial chemistry. Benzoylated D-glucose, D-galactose and L-quinovose DISAL glycosyl donors were synthesized in high yields and used under mild conditions to glycosylate methyl saphenate and 2-hydroxyphenazine. The glycosides were screened for biological activity and one compound showed inhibitory activity towards topoisomerase II.
Sigma-Aldrich
3,5-Dinitrosalicylic acid, 98%