Merck
  • Galactosylceramide containing omega-amino-fatty acids: preparation, characterization, and sulfotransferase acceptor.
Galactosylceramide containing omega-amino-fatty acids: preparation, characterization, and sulfotransferase acceptor.
Journal of lipid research (1992-08-01)
K Kamio, S Gasa, A Makita
ABSTRACT
For preparation of an affinity ligand, an N-fatty acyl moiety of galactosylceramide (GalCer) was chemically replaced with omega-amino-fatty acid including amino-n-hexanoic acid or amino-n-dodecanoic acid to obtain omega-aminoGalCer. For the synthesis of the compound, galactosylsphingosine (GalSph) was coupled with N-trifluoroacetyl omega-amino-fatty acid which was prepared by a reaction with S-ethyltrifluorothioacetate. After removal of the N-trifluoroacetyl group in a mild alkaline solution, in which an N-fatty acyl group was retained, aminoGalCer composed of an N-hexanoyl or an N-dodecanoyl group was obtained with an overall yield of 90%. Their chemical structures were confirmed by proton nuclear magnetic resonance and fast atom bombardment-mass spectrometries. These aminoGalCers and GalSph as well as immobilized aminoGalCer were sulfated by a glycolipid sulfotransferase from rat kidney. Furthermore, immobilized aminoGalCer on gel matrix was used for affinity chromatography of the sulfotransferase, resulting in an excellent increase in the purification (14,000-fold) with a recovery rate of 40%.
Sigma-Aldrich
12-Aminododecanoic acid, 95%
Pricing and availability is not currently available.