Merck
  • Intracellular singlet oxygen generation by phagocytosing neutrophils in response to particles coated with a chemical trap.
Intracellular singlet oxygen generation by phagocytosing neutrophils in response to particles coated with a chemical trap.
The Journal of biological chemistry (1992-07-05)
M J Steinbeck, A U Khan, M J Karnovsky
ABSTRACT
To determine if singlet oxygen (O2(1 delta g)) is produced by neutrophils (PMNs) during the process of phagocytosis, glass beads were coated with a specific chemical trap for O2(1 delta g), 9,10-diphenylanthracene (DPA). Singlet oxygen, but not other reactive oxygen species, reacts rapidly with DPA at a rate of kr = 1.3 x 10(6) M-1 s-1 to form a stable product, DPA-endoperoxide (Corey, E. J., and Taylor, W. C. (1964) J. Am. Chem. Soc. 86, 3881-3882; Wasserman, H. H., Scheffer, J. R., and Cooper, J. L. (1972) J. Am. Chem. Soc. 94, 4991-4996; Turro, N. J., Chow, M.-F., and Rigaudy, J. (1981) J. Am. Chem. Soc. 103, 7218-7224). The production of DPA-endoperoxide was determined by ultraviolet spectroscopy as a decrease in DPA absorbance at 355 nm. The absorbance of DPA was normalized to the absorbance of perylene, which was included in the coating on the beads as a nonreactive, internal standard. In the present study, DPA- and perylene-coated beads were initially allowed to adhere to fibronectin-coated coverslips. PMNs were then added to the bead-coated coverslips and allowed to adhere and phagocytose the beads for 1 h at 37 degrees C. In some experiments, 4B-phorbol-12-myristate-13-acetate (PMA) (1 ng/2.5 x 10(7) cells/ml), a known activator of the PMN NADPH-oxidase, was added as a co-stimulant. The amount of O2(1 delta g) produced by phagocytically stimulated PMNs was calculated to be 11.3 +/- 4.9 nmol of O2(1 delta g)/1.25 x 10(6) cells. Low dose PMA co-stimulation increased the production of O2(1 delta g) to 14.1 +/- 4.1 nmol/1.25 x 10(6) cells. Averaged together these amounts represent approximately 19 +/- 5.0% of the total oxygen consumed by PMNs in response to DPA- and perylene-coated beads. The specificity of the DPA reaction with O2(1 delta g) was confirmed by warming to 120 degrees C, which releases O2(1 delta g) from the DPA-endoperoxide, regenerating the parent DPA compound (Wasserman et al., 1972; Turro et al., 1981) and the absorbance at 355 nm. In addition, beta-carotene, an avid quencher of O2(1 delta g), was included in the coating of some bead preparations; assays in which these beads were used showed no change in the absorbance at 355 nm. Singlet oxygen production by myeloperoxidase was also measured using the coated bead assay and the results suggest that this is a major pathway by which singlet oxygen is generated in phagocytically stimulated PMNs.(ABSTRACT TRUNCATED AT 400 WORDS)
Sigma-Aldrich
9,10-Diphenylanthracene, 97%
Pricing and availability is not currently available.
Supelco
9,10-Diphenylanthracene, analytical standard
Pricing and availability is not currently available.