Merck
  • Synthesis of mesoporous silica with embedded nickel nanoparticles for catalyst applications.
Synthesis of mesoporous silica with embedded nickel nanoparticles for catalyst applications.
Journal of nanoscience and nanotechnology (2003-08-12)
E R Leite, N L Carreño, E Longo, A Valentini, L F Probst
ABSTRACT
Here we describe a new route for the synthesis of nanometric Ni particles embedded in a mesoporous silica material with excellent potential for catalytic applications. Mesoporous silica with a surface area in the range of 202-280 m2/g, with narrow pore size distribution and Ni nanoparticles (particles in the range of 3-41 nm) were obtained in a direct process. A different approach was adopted to process such a nanocomposite. This new approach is based on the formation of a polymer with the silicon oxianion and nickel cation chelated to the macromolecule structure and on the control of the pyrolysis step. The CO/CO2 atmosphere resulting from the pyrolysis of the organic material promotes the reduction of the Ni citrate.
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Pricing and availability is not currently available.