- Paraganglioma of the cervical sympathetic trunk.
Paraganglioma of the cervical sympathetic trunk.
The Annals of otology, rhinology, and laryngology (2002-12-25)
Manfred T Birchler, Klara Landau, Philip T Went, Sandro J Stoeckli
PMID12498369
ABSTRACT
We report the case of a 17-year-old man who presented with left-sided Horner's syndrome. Magnetic resonance imaging revealed a spindle-shaped cervical tumor in the left paravertebral space. During operation, a tumor originating from the left sympathetic trunk was found. The histopathologic analysis showed a sympathetic paraganglioma. The sympathetic trunk is an extremely rare location for a cervical paraganglioma; only a few cases have been reported in the literature.