Merck
  • Structure of the arabinogalactan from gum tragacanth (Astralagus gummifer).
Structure of the arabinogalactan from gum tragacanth (Astralagus gummifer).
Carbohydrate research (2002-11-09)
Cesar A Tischer, Marcello Iacomini, Philip A J Gorin
ABSTRACT
The polysaccharide obtained by ethanol precipitation from an aqueous solution of gum tragacanth contained arabinogalactan and tragacanthic acid, as well as starch ( approximately 0.6%). GC-MS, NMR, and ESI-MS analyses showed the structure of the arabinogalactan to be even more complex than previously determined, with core structures containing Arap, beta-Araf, and alpha-Galp units, as well as known terminal, and 2-O- and 3-O-substituted alpha-Araf units. Analysis was aided by examination of free, reducing oligosaccharides present in the gum. In addition to maltose, maltotriose, maltotetraose, and maltopentaose, the following were characterized: mixed alpha-Araf (1-->2)-alpha-Araf-(1-->4)-Ara and alpha-Araf-(1-->2)-alpha-Araf-(1-->5)-Ara, which correspond to the side chains of the arabinogalactan, beta-Galp-(1-->4)-beta-Galp-(1-->4)-beta-Galp-(1-->4)-Gal; and a mixture of beta-Galp-(1-->4)-beta-Galp-(1-->4)-Gal and beta-Glcp-(1-->4)-beta-Galp-(1-->4)-beta-Galp-(1-->4)-Gal, which did not resemble side-chain structures of the arabinogalactan. The latter are suggested to be related to tragacanthic acid, which has been previously found to contain beta-Galp nonreducing end-units.