Merck
  • 1,3-Dibromo-5,5-dimethylhydantoin (DBH) as oxidant and precipitant for drug identification according to PH. EUR Analytical methods of pharmacopoeias with DBH in respect of environmental and economical concern, Part 15(1).
1,3-Dibromo-5,5-dimethylhydantoin (DBH) as oxidant and precipitant for drug identification according to PH. EUR Analytical methods of pharmacopoeias with DBH in respect of environmental and economical concern, Part 15(1).
Die Pharmazie (2002-07-16)
M Hilp
ABSTRACT
Elemental bromine serves as oxidant for the identification of propylthiouracil, 2-thiouracil and sulphur according to PH. EUR. 2002. Phenol is identified according to PH. EUR. 2002 with bromine water as the sparingly water-soluble 2,4,4,6-tetrabromo-2,5-cyclohexadiene-1-one. These tests can be performed better using 1,3-dibromo-5,5-dimethylhydantoin (DBH).
Sigma-Aldrich
1,3-Dibromo-5,5-dimethylhydantoin, 98%