Merck
  • Determination of semivolatile compounds in Baltic herring (Clupea harengus membras) by supercritical fluid extraction-supercritical fluid chromatography-gas chromatography-mass spectrometry.
Determination of semivolatile compounds in Baltic herring (Clupea harengus membras) by supercritical fluid extraction-supercritical fluid chromatography-gas chromatography-mass spectrometry.
Journal of agricultural and food chemistry (2002-03-21)
Tarja Aro, Cato Brede, Pekka Manninen, Heikki Kallio
ABSTRACT
The on-line supercritical fluid extraction-supercritical fluid chromatography-gas chromatography method was applied to the determination of volatile compounds of raw and baked Baltic herring (Clupea harengus membras). The analytes were extracted with supercritical carbon dioxide at 45 degrees C and 10 MPa pressure. After extraction, the volatiles and coeluted lipids were separated on-line using supercritical fluid chromatography and the volatile fraction was introduced directly into a gas chromatograph. In all, 30 compounds were identified from fish samples with mass spectrometry. The most abundant compounds in the fresh Baltic herrings were heptadecane and 1-heptadecene. When the fish were stored for 3-6 days at 6 degrees C, the total peak area of the volatiles began to increase and the proportions of short chain acids (acetic, propanoic, 2-methylpropanoic, and 3-methylbutanoic) also increased. After 8-9 days of storage, 3-methylbutanoic acid comprised about 36 and 40% of all volatiles in raw and baked herring, respectively.
Sigma-Aldrich
Heptadecane, 99%
Pricing and availability is not currently available.
Supelco
Heptadecane, analytical standard
Pricing and availability is not currently available.