Merck
  • Performance of a proposed determinative method for p-TSA in rainbow trout fillet tissue and bridging the proposed method with a method for total chloramine-T residues in rainbow trout fillet tissue.
Performance of a proposed determinative method for p-TSA in rainbow trout fillet tissue and bridging the proposed method with a method for total chloramine-T residues in rainbow trout fillet tissue.
Journal of AOAC International (2001-10-17)
J R Meinertz, G R Stehly, W H Gingerich, S L Greseth
ABSTRACT
Chloramine-T is an effective drug for controlling fish mortality caused by bacterial gill disease. As part of the data required for approval of chloramine-T use in aquaculture, depletion of the chloramine-T marker residue (para-toluenesulfonamide; p-TSA) from edible fillet tissue of fish must be characterized. Declaration of p-TSA as the marker residue for chloramine-T in rainbow trout was based on total residue depletion studies using a method that used time consuming and cumbersome techniques. A simple and robust method recently developed is being proposed as a determinative method for p-TSA in fish fillet tissue. The proposed determinative method was evaluated by comparing accuracy and precision data with U.S. Food and Drug Administration criteria and by bridging the method to the former method for chloramine-T residues. The method accuracy and precision fulfilled the criteria for determinative methods; accuracy was 92.6, 93.4, and 94.6% with samples fortified at 0.5X, 1X, and 2X the expected 1000 ng/g tolerance limit for p-TSA, respectively. Method precision with tissue containing incurred p-TSA at a nominal concentration of 1000 ng/g ranged from 0.80 to 8.4%. The proposed determinative method was successfully bridged with the former method. The concentrations of p-TSA developed with the proposed method were not statistically different at p < 0.05 from p-TSA concentrations developed with the former method.
Sigma-Aldrich
p-Toluenesulfonamide, reagent grade, 97%
Sigma-Aldrich
p-Toluenesulfonamide, ReagentPlus®, ≥99%