Merck
  • Fate of seven pesticides in an aerobic aquifer studied in column experiments.
Fate of seven pesticides in an aerobic aquifer studied in column experiments.
Chemosphere (2000-11-01)
N Tuxen, P L Tüchsen, K Rügge, H J Albrechtsen, P L Bjerg
ABSTRACT
The fate of selected pesticides (bentazone, isoproturon, DNOC, MCPP, dichlorprop and 2,4-D) and a metabolite (2,6-dichlorobenzamide (BAM)) was investigated under aerobic conditions in column experiments using aquifer material and low concentrations of pesticides (approximately 25 microg/l). A solute transport model accounting for kinetic sorption and degradation was used to estimate sorption and degradation parameters. Isoproturon and DNOC were significantly retarded by sorption, whereas the retardation of the phenoxy acids (MCPP, 2,4-D and dichlorprop), BAM and bentazone was very low. After lag periods of 16-33 days for the phenoxy acids and 80 days for DNOC, these pesticides were degraded quickly with 0.-order rate constants of 1.3-2.6 microg/l/day. None of the most probable degradation products were detected.