Merck
  • Antiplasmodial compounds from the wood-decayed fungus Xylaria sp. BCC 1067.
Antiplasmodial compounds from the wood-decayed fungus Xylaria sp. BCC 1067.
Planta medica (2000-07-26)
M Isaka, A Jaturapat, W Kladwang, J Punya, Y Lertwerawat, M Tanticharoen, Y Thebtaranonth
ABSTRACT
Bioassay-guided fractionation of the extract from the wood-decayed fungus Xylaria sp. BCC 1067 led to the isolation of five antiplasmodial compounds, (-)-depudecin, (+)-phaseolinone, (+)-phomenone, 19,20-epoxycytochalasin Q, and (E)-methyl 3-(4-methoxyphenoxy)propenoate. These structures were elucidated using spectroscopic methods, especially NMR analysis.