Merck
  • Endothelin alters the reactivity of vasa vasorum: mechanisms and implications for conduit vessel physiology and pathophysiology.
Endothelin alters the reactivity of vasa vasorum: mechanisms and implications for conduit vessel physiology and pathophysiology.
British journal of pharmacology (1999-12-01)
R Scotland, P Vallance, A Ahluwalia
ABSTRACT
1 The walls of certain large blood vessels are nourished by the vasa vasorum, a network of microvessels that penetrate the adventitia and media of the vessel wall. The purpose of this study was to characterize endothelin-1 (ET-1)-mediated contraction of vasa and to investigate whether threshold concentrations of ET-1 alters the sensitivity to constrictors. Arterial vasa were dissected from the walls of porcine thoracic aorta and mounted in a tension myograph. 2 ET-1 and ETB-selective agonist, sarafotoxin 6c (S6c), produced concentration-dependent contraction. ETA receptor antagonist, BQ123 (10 microM), caused a biphasic rightward shift of ET-1 response curves. ETB receptor antagonist, BQ788 (1 microM), produced a rightward shift of response curves to ET-1 and S6c of 5- and 80 fold respectively. 3 ET-1 responses were abolished in Ca2+-free PSS but unaffected by selective depletion of intracellular Ca2+ stores. Nifedipine (10 microM), an L-type Ca2+ channel blocker, attenuated ET-1 responses by 44%. Inhibition of receptor-operated Ca2+ channels or non-selective cation entry using SKF 96365 (30 microM) and Ni2+ (1 mM) respectively, attenuated ET-1 contractions by 60%. 4 ET-1 (1-3 nM) enhanced responses to noradrenaline (NA) (4 fold) but not to thromboxane A2-mimetic, whilst K+ (10-20 mM) sensitized vasa to both types of constrictor. 5 Therefore, ET-1-induced contraction of isolated vasa is mediated by ETA and ETB receptors and involves Ca2+ influx through L-type and non-L-type Ca2+ channels. Furthermore elevation of basal tone of vasa vasorum alters the profile of contractile reactivity. These results suggest that ET-1 may be an important regulator of vasa vasorum reactivity.
Sigma-Aldrich
9,11-Dideoxy-9α,11α-epoxymethanoprostaglandin F