Merck
  • A novel sodium-hydrogen exchanger isoform-1 inhibitor, zoniporide, reduces ischemic myocardial injury in vitro and in vivo.
A novel sodium-hydrogen exchanger isoform-1 inhibitor, zoniporide, reduces ischemic myocardial injury in vitro and in vivo.
The Journal of pharmacology and experimental therapeutics (2001-03-22)
D R Knight, A H Smith, D M Flynn, J T MacAndrew, S S Ellery, J X Kong, R B Marala, R T Wester, A Guzman-Perez, R J Hill, W P Magee, W R Tracey
ABSTRACT
The cardioprotective efficacy of zoniporide (CP-597,396), a novel, potent, and selective inhibitor of the sodium-hydrogen exchanger isoform 1 (NHE-1), was evaluated both in vitro and in vivo using rabbit models of myocardial ischemia-reperfusion injury. In these models, myocardial injury was elicited with 30 min of regional ischemia and 120 min of reperfusion. Zoniporide elicited a concentration-dependent reduction in infarct size (EC(50) of 0.25 nM) in the isolated heart (Langendorff) and reduced infarct size by 83% (50 nM). This compound was 2.5- to 20-fold more potent than either eniporide or cariporide (EC(50) of 0.69 and 5.11 nM, respectively), and reduced infarct size to a greater extent than eniporide (58% reduction in infarct size). In open-chest, anesthetized rabbits, zoniporide also elicited a dose-dependent reduction in infarct size (ED(50) of 0.45 mg/kg/h) and inhibited NHE-1-mediated platelet swelling (maximum inhibition 93%). Furthermore, zoniporide did not cause any in vivo hemodynamic (mean arterial pressure, heart rate, rate pressure product) changes. Zoniporide represents a novel class of potent NHE-1 inhibitors with potential utility for providing clinical cardioprotection.
Sigma-Aldrich
Zoniporide hydrochloride hydrate, ≥98% (HPLC)