Merck
  • Modulation of TLR signaling by multiple MyD88-interacting partners including leucine-rich repeat Fli-I-interacting proteins.
Modulation of TLR signaling by multiple MyD88-interacting partners including leucine-rich repeat Fli-I-interacting proteins.
Journal of immunology (Baltimore, Md. : 1950) (2009-03-07)
Penggao Dai, Sun Yong Jeong, Yanbao Yu, Taohua Leng, Weidong Wu, Ling Xie, Xian Chen
ABSTRACT
Emerging evidences suggest TLR-mediated signaling is tightly regulated by a specific chain of intracellular protein-protein interactions, some of which are yet to be identified. Previously we utilized a dual-tagging quantitative proteomics approach to uncover MyD88 interactions in LPS-stimulated cells and described the function of Fliih, a leucine-rich repeat (LRR) protein that negatively regulates NF-kappaB activity. Here we characterize two distinct LRR-binding MyD88 interactors, LRRFIP2 and Flap-1, and found that both are positive regulators of NF-kappaB activity. Upon LPS stimulation, LRRFIP2 was also found to positively regulate cytokine production in macrophages, suggesting a functional role in TLR4-mediated inflammatory response. Furthermore, we observed that immediately following LPS stimulation both LRRFIP2 and Flap-1 compete with Fliih for interacting with MyD88 to activate the signaling. By using a novel multiplex quantitative proteomic approach, we found that at endogenous levels these positive and negative regulators interact with MyD88 in a timely and orderly manner to differentially mediate the NF-kappaB activity through the course of signaling from initiation to prolongation, and to repression. Based on these data, we describe a mechanistic model in which selective modulation of TLR signaling is achieved by temporal and dynamic interactions of MyD88 with its regulators.
Product Description
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
iTRAQ® Reagents Multiplex Kit