Merck
  • The Flt3 receptor tyrosine kinase collaborates with NUP98-HOX fusions in acute myeloid leukemia.
The Flt3 receptor tyrosine kinase collaborates with NUP98-HOX fusions in acute myeloid leukemia.
Blood (2006-07-25)
Lars Palmqvist, Bob Argiropoulos, Nicolas Pineault, Carolina Abramovich, Laura M Sly, Gerald Krystal, Adrian Wan, R Keith Humphries
ABSTRACT
In leukemogenesis, several genetic changes conferring a proliferative and/or survival advantage to hematopoietic progenitor cells in addition to a block in differentiation are required. Here, we demonstrate that overexpression of the wild-type (wt) Flt3 receptor tyrosine kinase collaborates with NUP98-HOX fusions (NUP98-HOXA10 and NUP98-HOXD13) to induce aggressive acute myeloid leukemia (AML). We used a mouse transplantation model to show their synergism in cotransduced bone marrow cells as well as in a cellular model of leukemic progression. Furthermore, our data support the finding that Meis1 overexpression leads to marked elevation in Flt3 transcription and extend it to the context of NUP98-HOX-induced leukemia. Together, these results support a multistep model where the synergism between NUP98-HOX and wt-Flt3 is the result of the ability of Flt3 to increase proliferation of myeloid progenitors blocked in differentiation by NUP98-HOX fusions and reveal a direct role for wt-Flt3 in the pathobiology of AML. Given the similarities in the leukemogenic role of native HOX and NUP98-fused HOX genes, our results underscore the clinical significance of the recurrent co-overexpression of wt-FLT3 and HOX in human leukemia and suggest that specific FLT3 inhibitors could be useful in treatment of HOX-induced AML or acute lymphoblastic leukemia (ALL).
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution