Merck
Serine protease inhibitor A3 in atherosclerosis and aneurysm disease.
International journal of molecular medicine (2012-05-15)
Dick Wågsäter, Daniel Johansson, Vincent Fontaine, Emina Vorkapic, Alexandra Bäcklund, Anton Razuvaev, Mikko I Mäyränpää, Charlotta Hjerpe, Kenneth Caidahl, Anders Hamsten, Anders Franco-Cereceda, Johannes Wilbertz, Jesper Swedenborg, Xinghua Zhou, Per Eriksson
ABSTRACT
Remodeling of extracellular matrix (ECM) plays an important role in both atherosclerosis and aneurysm disease. Serine protease inhibitor A3 (serpinA3) is an inhibitor of several proteases such as elastase, cathepsin G and chymase derived from mast cells and neutrophils. In this study, we investigated the putative role of serpinA3 in atherosclerosis and aneurysm formation. SerpinA3 was expressed in endothelial cells and medial smooth muscle cells in human atherosclerotic lesions and a 14-fold increased expression of serpinA3n mRNA was found in lesions from Apoe-/- mice compared to lesion-free vessels. In contrast, decreased mRNA expression (-80%) of serpinA3 was found in biopsies of human abdominal aortic aneurysm (AAA) compared to non-dilated aortas. Overexpression of serpinA3n in transgenic mice did not influence the development of atherosclerosis or CaCl2-induced aneurysm formation. In situ zymography analysis showed that the transgenic mice had lower cathepsin G and elastase activity, and more elastin in the aortas compared to wild-type mice, which could indicate a more stable aortic phenotype. Differential vascular expression of serpinA3 is clearly associated with human atherosclerosis and AAA but serpinA3 had no major effect on experimentally induced atherosclerosis or AAA development in mouse. However, serpinA3 may be involved in a phenotypic stabilization of the aorta.
Sigma-Aldrich
Cathepsin G from human leukocytes, lyophilized powder, ≥60 units/mg protein (Bradford)
Pricing and availability is not currently available.