- Label free fluorescence turn-on detection of polynucleotide kinase activity with a perylene probe.
Label free fluorescence turn-on detection of polynucleotide kinase activity with a perylene probe.
Chemical communications (Cambridge, England) (2012-07-13)
Huping Jiao, Bin Wang, Jian Chen, Dongli Liao, Wenying Li, Cong Yu
PMID22786467
ABSTRACT
A single stranded oligonucleotide could induce aggregation of a perylene probe, the probe's monomer fluorescence was efficiently quenched. However, when the oligonucleotide was 5'-phosphorylated by polynucleotide kinase, it could be very efficiently degraded by lambda exonuclease, probe monomers were released, and a turn on fluorescence signal was detected.
Sigma-Aldrich
Polynucleotide Kinase from T4-infected Escherichia coli, 10 units/μL, buffered aqueous glycerol solution