Merck
  • Effects of konjac glucomannan on putative risk factors for colon carcinogenesis in rats fed a high-fat diet.
Effects of konjac glucomannan on putative risk factors for colon carcinogenesis in rats fed a high-fat diet.
Journal of agricultural and food chemistry (2011-01-07)
Wen-Tzu Wu, Hsiao-Ling Chen
ABSTRACT
The aim of this study was to determine effects of konjac glucomannan (KGM) in a high fat corn oil diet on risk factors of colon carcinogenesis, that is, fecal β-glucuronidase, mucinase, and bile acids, and on preventive factors, that is, fecal microflora and cecal short-chain fatty acids (SCFAs). Sprague-Dawley rats (n = 8 animals per group) were fed a normal-fat fiber-free (5% corn oil, w/w) or high-fat (25% corn oil, w/w) diet containing no fiber, KGM (5%, w/w), or inulin (5%, w/w, as a prebiotic control) for 4 weeks. Results indicated that the high-fat fiber-free diet significantly elevated the fecal β-glucuronidase and mucinase activities and total bile acid concentration and decreased cecal SCFA contents, as compared with its normal-fat counterpart. The incorporation of KGM, as well as inulin, into the high-fat fiber-free diet beneficially reduced the fecal β-glucuronidase and mucinase activities and lithocholic acid (secondary bile acid) concentration. Although KGM elevated the daily fecal total bile acid excretion, the change was due to the primary, instead of the secondary, bile acids. In addition, KGM beneficially promoted the daily fecal excretion of bifidobacteria and lactobacilli and cecal SCFA contents, as compared with the high-fat fiber-free diet. Therefore, the present study suggests that KGM potentially attenuated the high fat-induced risk in colon carcinogenesis.
Sigma-Aldrich
Hyaluronidase from Streptomyces hyalurolyticus
Pricing and availability is not currently available.