Merck
  • Development and characterization of an enzymatic method for the rapid determination of gamma hydroxybutyric acid.
Development and characterization of an enzymatic method for the rapid determination of gamma hydroxybutyric acid.
Chimia (2011-01-05)
Michel A Sciotti, Lara Hasan, Andre Scholer, Thomas M Jermann, Jakob M Weber, Daniel Gygax
ABSTRACT
Gamma hydroxybutyric acid (GHB) is a regulated therapeutic drug, which naturally occurs in mammalian brain tissues as an intermediate of the GABA (gamma aminobutyric acid) neurotransmitter metabolism. The increasing misuse of GHB as a narcotic or abusing drug in recent years calls for the development of a simple and rapid screening method as an alternative to the currently available, technically demanding diagnostic methods. We have developed a rapid enzymatic assay based on the GHB dehydrogenase of Ralstonia eutropha. The enzyme is expressed as a recombinant protein in Escherichia coli and characterized in terms of reaction mechanism and kinetic parameters for the catalysis of conversion of GHB into succinic semialdehyde (SSA). The concomitant NADH production enables spectrophotometric monitoring of the reaction and the quantification of GHB in physiological fluids depending on initial velocities. We have tested a panel of twelve serum and urine samples containing GHB concentrations from 0.0 to 2.1 mmol/L. GHB dehydrogenase activity obeys a non classical bi bi ping pong mechanism exhibiting substrate inhibition by NAD+. With an optimal NAD+ concentration of 3.7 mmol/L in the reaction, the enzyme yields a K(M) of 1.0 mmol/L for GHB and a Vmax of 3.37 mmol/min/mg. The assay shows a linear standard curve from 0.1 to at least 1 mmol/L of GHB. Spiking experiments result in mean recoveries of 92% for urine and 114% for serum, respectively. The comparison to an ion chromatographic reference method exhibits a mean difference of 10% divergence from the target values in urine and 9% in serum, respectively.
Sigma-Aldrich
β-Hydroxybutyrate Dehydrogenase from Pseudomonas lemoignei, lyophilized powder, ≥200 units/mg protein
Pricing and availability is not currently available.
Sigma-Aldrich
β-Hydroxybutyrate Dehydrogenase from Rhodopseudomonas sphaeroides, Type V, lyophilized powder, 250-750 units/mg protein
Pricing and availability is not currently available.