Merck
  • Encapsulation of yeast displaying glucose oxidase on their surface in graphene oxide hydrogel scaffolding and its bioactivation.
Encapsulation of yeast displaying glucose oxidase on their surface in graphene oxide hydrogel scaffolding and its bioactivation.
Chemical communications (Cambridge, England) (2012-11-06)
Karnit Bahartan, Jenny Gun, Sergey Sladkevich, Petr V Prikhodchenko, Ovadia Lev, Lital Alfonta
ABSTRACT
Yeast displaying glucose oxidase on their surface were encapsulated in a graphene oxide hydrogel. The ability of the modified yeast to reduce graphene oxide by glucose assimilation while maintaining viability was tested with time and deemed suitable for biofuel cell applications.
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type II, ≥10,000 units/g solid (without added oxygen)
Pricing and availability is not currently available.
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Pricing and availability is not currently available.
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type X-S, lyophilized powder, 100,000-250,000 units/g solid (without added oxygen)
Pricing and availability is not currently available.