Merck
  • Dietary lipid content influences the activity of lipogenic enzymes in the liver and on whole body delta13C values of Nile tilapia, Oreochromis niloticus (L.).
Dietary lipid content influences the activity of lipogenic enzymes in the liver and on whole body delta13C values of Nile tilapia, Oreochromis niloticus (L.).
Isotopes in environmental and health studies (2004-09-17)
Julia Gaye-Siessegger, Ulfert Focken, Hansjörg Abel, Klaus Becker
ABSTRACT
The use of stable isotope techniques for the reconstruction of diets has increased over the last decade. However, isotopic ratios in an animal are not only affected by the composition of the feed, but also by the amount of feed consumed. An uncertainty of up to 1 per thousand for both delta13C and delta15N values has been observed when the feeding level is unknown. This may have substantial effects on the results of back-calculation. As the feeding level of animals is unknown in nature, an additional indicator for their nutritional status is needed. High feeding levels and a consequent surfeit of dietary energy lead to the synthesis of lipids. In order to test whether the level of lipogenesis could be used as an indicator, Nile tilapia (Oreochromis niloticus) were fed four isonitrogenous and isoenergetic wheat-based semi-synthetic diets with different lipid contents (2.0 %, 4.5 %, 9.5 % and 13.3 %) for eight weeks. Body composition, gross energy content and delta13C values in the lipids and the lipid-free material were determined in diets and fish bodies. The livers of three fish per feeding group were assayed for the activity of two lipogenic enzymes, ATP-citrate lyase and malic enzyme. There was a strong negative correlation between delta13C values in the lipids of the individual fish and the apparent lipid conversion. The activities of lipogenic enzymes decreased with rising lipid content in the diet. The delta13C values in the lipids decreased significantly with increasing specific activity for both enzymes. In this experiment where lipogenesis was influenced by the composition of the diet, it was possible to determine the exact value for the trophic shift in relation to the enzyme activities. Further experiments to investigate the use of enzyme activities in situations where the feeding level of an animal is unknown are recommended.
Sigma-Aldrich
Citrate Lyase from Klebsiella pneumoniae, lyophilized powder, ≥0.20 unit/mg solid
Pricing and availability is not currently available.